missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbyl 2,6-Dibutyrate 98.0+%, TCI America™

Catalog No. A12051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12051G Supplier TCI America Supplier No. A12051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4337-04-6
Molecular Formula C14H20O8
Molecular Weight (g/mol) 316.31
MDL Number MFCD00144561
InChI Key BFXWCTCPRAYDEB-UHFFFAOYNA-N
Synonym 2,6-Di-O-butyryl-L-ascorbic Acid
PubChem CID 124632456
IUPAC Name [(2R)-2-[(2S)-4-butanoyloxy-3-hydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] butanoate
SMILES CCCC(=O)OCC(C1C(=C(C(=O)O1)OC(=O)CCC)O)O

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 316.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Ascorbyl 2,6-Dibutyrate
TSCA No
Recommended Storage Refrigerator