missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Methyl Ester Dihydrochloride 98.0+%, TCI America™

Catalog No. A20175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20175G 5 g
A201725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20175G Supplier TCI America Supplier No. A20175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26340-89-6
Molecular Formula C7H18Cl2N4O2
Molecular Weight (g/mol) 261.147
MDL Number MFCD00038948
InChI Key XQYZOBNLCUAXLF-XRIGFGBMSA-N
Synonym l-arginine methyl ester dihydrochloride, h-arg-ome.2hcl, methyl l-argininate dihydrochloride, h-arg-ome 2hcl, l-arginine, methyl ester, dihydrochloride, s-methyl 2-amino-5-guanidinopentanoate dihydrochloride, methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride, l-arginine, methyl ester, hydrochloride 1:2, h-arg-ome?hcl, h-arg-ome 2 hcl
PubChem CID 117776
IUPAC Name methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
SMILES COC(=O)C(CCCN=C(N)N)N.Cl.Cl

Specifications

Melting Point 193°C
Color White
Quantity 5 g
Formula Weight 261.15
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine Methyl Ester Dihydrochloride