missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine methyl ester dihydrochloride, 98%

Catalog No. AAL0595914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0595914 25 g
AAL0595906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0595914 Supplier Thermo Scientific Chemicals Supplier No. L0595914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26340-89-6 | C7H18Cl2N4O2 | 261.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26340-89-6
Molecular Formula C7H18Cl2N4O2
Molecular Weight (g/mol) 261.147
MDL Number MFCD00038948
InChI Key XQYZOBNLCUAXLF-XRIGFGBMSA-N
PubChem CID 117776
IUPAC Name methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
SMILES COC(=O)C(CCCN=C(N)N)N.Cl.Cl

Specifications

Melting Point ∽190°C (decomposition)
Beilstein 4159929
Synonym l-arginine methyl ester dihydrochloride, h-arg-ome.2hcl, methyl l-argininate dihydrochloride, h-arg-ome 2hcl, l-arginine, methyl ester, dihydrochloride, s-methyl 2-amino-5-guanidinopentanoate dihydrochloride, methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride, l-arginine, methyl ester, hydrochloride 1:2, h-arg-ome?hcl, h-arg-ome 2 hcl
Quantity 25 g
Formula Weight 261.15
Optical Rotation +21° (c=2.5 in Methanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Arginine methyl ester dihydrochloride
EINECSNumber 247-622-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only