missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Arginine, Hydrochloride, Calbiochem™,

$133.00 - $704.00

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18100350GM
View Documents Promotion Details
MilliporeSigma™
18100350GM
50 g Glass Bottle
Each for $133.00
Only null left
 
1810031KG
View Documents Promotion Details
MilliporeSigma™
1810031KG
1 kg Glass Bottle
Each for $704.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1119-34-2
210.662
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C6H15ClN4O2
KWTQSFXGGICVPE-WCCKRBBISA-N
66250
C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

1119-34-2
C6H15ClN4O2
KWTQSFXGGICVPE-WCCKRBBISA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
66250
Solid
White
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
C(CC(C(=O)O)N)CN=C(N)N.Cl
210.662
≥99% (Perchloric Acid Titration)
Videos
SDS
Documents

Documents