missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Arginine, Hydrochloride, Calbiochem™,

Supplier:  MilliporeSigma™ 1810031KG

Quantity 1 kg
Packaging Glass Bottle

 View more versions of this product

Catalog No. 1810031KG


Only null left

Description

Description

Specifications

Chemical Identifiers

1119-34-2
210.662
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C6H15ClN4O2
KWTQSFXGGICVPE-WCCKRBBISA-N
66250
C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

1119-34-2
C6H15ClN4O2
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
C(CC(C(=O)O)N)CN=C(N)N.Cl
1 kg
66250
Solid
White
Glass Bottle
KWTQSFXGGICVPE-WCCKRBBISA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
210.662
≥99% (Perchloric Acid Titration)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions