missing translation for 'onlineSavingsMsg'
Learn More

L-Alloisoleucine 95.0+%, TCI America™

Catalog No. A2214200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2214200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2214200MG Supplier TCI America Supplier No. A2214200MG

Chemical Identifiers

CAS 1509-34-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066446
InChI Key AGPKZVBTJJNPAG-BJNJOQLTNA-N
Synonym l-allo-isoleucine, l-alloisoleucine, alloisoleucine, 2s,3r-2-amino-3-methylpentanoic acid, allo-isoleucine, allo-l-isoleucine, h-allo-ile-oh, 3r-ls-isoleucine, isoleucine, allo, isoleucine, threo
PubChem CID 99288
ChEBI CHEBI:43433
IUPAC Name (2S,3R)-2-amino-3-methylpentanoic acid
SMILES CC[C@@H](C)[C@H](N)C(O)=O

Specifications

Melting Point 272°C
Color White
Quantity 200 mg
Formula Weight 131.18
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alloisoleucine
EINECSNumber (9)-1560
RTECSNumber BA2933000
TSCA No