missing translation for 'onlineSavingsMsg'
Learn More

L-allo-Isoleucine, 99%, 99% ee

Catalog No. AC399491000
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399491000 100 mg
AC399490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399491000 Supplier Thermo Scientific Chemicals Supplier No. 399491000

CAS: 1509-34-8 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1509-34-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066446
InChI Key AGPKZVBTJJNPAG-BJNJOQLTNA-N
Synonym l-allo-isoleucine, l-alloisoleucine, alloisoleucine, 2s,3r-2-amino-3-methylpentanoic acid, allo-isoleucine, allo-l-isoleucine, h-allo-ile-oh, 3r-ls-isoleucine, isoleucine, allo, isoleucine, threo
PubChem CID 99288
ChEBI CHEBI:43433
IUPAC Name (2S,3R)-2-amino-3-methylpentanoic acid
SMILES CC[C@@H](C)[C@H](N)C(O)=O

Specifications

Assay Percent Range 99%
Linear Formula CH3CH2CH(CH3)CH(NH2)COOH
Quantity 100 mg
Formula Weight 131.17
Chemical Name or Material L-allo-Isoleucine, 99%

RUO – Research Use Only