missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine 99.0+%, TCI America™
$47.46 - $197.46
Chemical Identifiers
| CAS | 56-41-7 |
|---|---|
| Molecular Formula | C3H7NO2 |
| Molecular Weight (g/mol) | 89.09 |
| MDL Number | MFCD00064410 |
| InChI Key | QNAYBMKLOCPYGJ-REOHCLBHSA-N |
| Synonym | l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid |
| PubChem CID | 5950 |
| ChEBI | CHEBI:16977 |
| IUPAC Name | (2S)-2-aminopropanoic acid |
| SMILES | C[C@H](N)C(O)=O |
Chemical Identifiers
| C3H7NO2 | |
| MFCD00064410 | |
| l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
| CHEBI:16977 | |
| C[C@H](N)C(O)=O |
Specifications
| 315°C | |
| White | |
| MFCD00064410 | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| (2S)-2-aminopropanoic acid | |
| 5950 | |
| 89.09 | |
| Crystalline Powder |
| 56-41-7 | |
| C3H7NO2 | |
| l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
| C[C@H](N)C(O)=O | |
| 89.09 | |
| CHEBI:16977 | |
| ≥99.0% (T) | |
| L-Alanine |
Safety and Handling
EINECSNumber : (9)-1553
RTECSNumber : AY2990000
TSCA : Yes