missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine 99.0+%, TCI America™

Supplier:  TCI America A0179250G

Quantity 250 g

 View more versions of this product

Catalog No. A0179250G


Only null left

Description

Description

Specifications

Chemical Identifiers

56-41-7
89.09
QNAYBMKLOCPYGJ-REOHCLBHSA-N
5950
(2S)-2-aminopropanoic acid
C3H7NO2
MFCD00064410
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
CHEBI:16977
C[C@H](N)C(O)=O

Specifications

315°C
56-41-7
C3H7NO2
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
C[C@H](N)C(O)=O
89.09
5950
89.09
Crystalline Powder
L-Alanine
White
MFCD00064410
QNAYBMKLOCPYGJ-REOHCLBHSA-N
(2S)-2-aminopropanoic acid
250 g
CHEBI:16977
≥99.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1553

RTECSNumber : AY2990000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions