missing translation for 'onlineSavingsMsg'
Learn More

L(+)-2-Aminobutyric acid, 98%

Numéro de catalogue. AC215380010
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC215380010 1 g Glass bottle
AC215380050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC215380010 Fournisseur Thermo Scientific Chemicals Code fournisseur 215380010
Il en reste null

CAS: 1492-24-6 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1492-24-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00064415
InChI Key QWCKQJZIFLGMSD-VKHMYHEASA-N
Synonym l-2-aminobutyric acid, s-2-aminobutanoic acid, 2s-2-aminobutanoic acid, h-abu-oh, l-alpha-aminobutyric acid, s-+-2-aminobutyric acid, l-+-2-aminobutyric acid, --2-aminobutyric acid, s-2-aminobutyric acid, s-2-amino-butyric acid
PubChem CID 80283
ChEBI CHEBI:35619
IUPAC Name (2S)-2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Spécifications

Melting Point >300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5CH(NH2)CO2H
Quantity 1 g
Beilstein 04, 408
Merck Index 10, 430
Specific Rotation + 21.6
Solubility Information Solubility in water: 22.7g/100mL (22 c)
Formula Weight 103.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-2-Aminobutyric acid, 98%
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 216-083-3
TSCA TSCA

RUO – Research Use Only