missing translation for 'onlineSavingsMsg'
Learn More

Charles River Laboratories Inc KTA-Kinetic Turbidimetric LAL Reagent, 6-pack of 50-test vials, 5.2 mL per vial

Numéro de catalogue. NC3585260
Encompass

The Charles River Laboratories KTA-Kinetic Turbidimetric LAL Reagent is an FDA-licensed assay designed for quantitative detection of bacterial endotoxins. This reagent is suitable for both kinetic turbidimetric and gel-clot analyses, ensuring accurate results even in complex samples due to its buffered formulation. It requires a microplate reader and endotoxin-measuring software for operation, which enhances throughput and offers reliable quantitative results.

  • Assay type: Kinetic turbidimetric LAL, Gel-Clot
  • Application: Bacterial endotoxin detection, quantitative
  • Sensitivity: 0.015 EU/mL
  • Pack size: 6 vials
  • Interference resistance: Significant
  • Regulatory status: FDA-licensed for product release testing
  • Compatibility: Microplate reader or multi-tube reader with endotoxin-measuring software
  • Correlation: Direct correlation between kinetic and gel-clot methods
  • Reconstitution volume: 5.2 mL

Numéro de catalogue. NC3585260 Fournisseur Charles River Laboratories Inc Code fournisseur R15015PK

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Charles River Laboratories KTA-Kinetic Turbidimetric LAL Reagent is an FDA-licensed assay designed for quantitative detection of bacterial endotoxins. This reagent is suitable for both kinetic turbidimetric and gel-clot analyses, ensuring accurate results even in complex samples due to its buffered formulation. It requires a microplate reader and endotoxin-measuring software for operation, which enhances throughput and offers reliable quantitative results.

  • Assay type: Kinetic turbidimetric LAL, Gel-Clot
  • Application: Bacterial endotoxin detection, quantitative
  • Sensitivity: 0.015 EU/mL
  • Pack size: 6 vials
  • Interference resistance: Significant
  • Regulatory status: FDA-licensed for product release testing
  • Compatibility: Microplate reader or multi-tube reader with endotoxin-measuring software
  • Correlation: Direct correlation between kinetic and gel-clot methods
  • Reconstitution volume: 5.2 mL