missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KRTAP1-3 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA571738
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA571738 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA571738 Fournisseur Invitrogen™ Code fournisseur PA571738
Il en reste null

Rabbit Polyclonal Antibody

KRTAP1-3 Polyclonal Antibody for Western Blot, Flow

In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.

Spécifications

Antigen KRTAP1-3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KRTAP1-3
Gene Accession No. Q8IUG1
Gene Alias B2B; KAP1.2; KAP1.3; KAP1.6; KAP1.8; KAP1.8A; KAP1.8B; KAP1.9; keratin associated protein 1.6; keratin associated protein 1-3; Keratin-associated protein 1.8; keratin-associated protein 1.9; keratin-associated protein 1-3; KRATP1.9; KRTAP1.3; KRTAP1.8; KRTAP1-3
Gene Symbols KRTAP1-3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 88-117 amino acids from the Central region of human KRTAP1-3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81850
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats