Learn More
MilliporeSigma™ Supelco™ Kromasil™ Eternity™ C18 HPLC Column, 100 Å, 5 μm Particle Size
Kromasil™ Eternity™ and Eternity™XT can be used in a wider pH window than regular silica-based HPLC materials, giving more flexibility than ever when running analytical applications or optimizing for large-scale applications.
$2089.29 - $2089.29
Specifications
| Particle Size | 5 μm |
|---|---|
| Pore Size | 100 Å |
| Surface Area | 330 m2/g |
| Stationary Phase | C18 (octadecyl) |
| Product Line | Kromasil Eternity |
Description
For regular silica-based stationary phases, exposure to extreme pH (especially basic) will have a negative impact on the chemical stability and therefore column lifetime. However, the silica/organosilane surface of the Kromasil™ Eternity™ platform offers a chemical stability that will secure a long-lasting stationary phase, even under tough pH conditions and higher temperatures.Specifications
| 5 μm | |
| 330 m2/g | |
| Kromasil Eternity | |
| Reversed Phase |
| 100 Å | |
| C18 (octadecyl) | |
| C18 (octadecyl) Phase |
Kromasil and Eternity are trademarks of EKA Chemicals AB