Learn More
DWK Life Sciences Kimble™ Kontes™ Straight Bore Stopcock with PTFE Plug
Catalog No. K8210000002
Change view
Click to view available options
Plug Bore:
2 mm
Catalog No. Plug Bore
K8210000002 2 mm
1 options
Catalog No. K8210000002 Supplier DWK Life Sciences Supplier No. 8210000002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The more exaggerated taper of this stopcock permits easier use at higher-than-ambient temperatures.
  • With a 1:5 taper PTFE plug and a threaded pressure nut
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Type Stopcock Plug
Plug Bore 2 mm
Material Borosilicate Glass
Certifications/Compliance ASTM E438
Includes 1:5 taper PTFE plug and a threaded pressure nut