missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Amber 203 Serum Vials

Catalog No. 22268786
Change view
Click to view available options
Volume (Metric):
2 mL
Height (Metric):
40 mm
Diameter (Metric) Outer:
15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Height (Metric) Diameter (Metric) Outer
22268786 2 mL 40 mm 15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22268786 Supplier DWK Life Sciences Supplier No. 62413D2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ KONTES™ Amber 203 Serum Vials are autoclavable and ideally suited for handling and storage of reagents, vaccines, blood plasma, culture media, and chromatography samples. Made from amber 203 borosilicate glass with GPI aluminum seal finishes.

  • Autoclavable serum vials suited for handling, containment, and storage of reagents, vaccines, blood plasma, culture media, and chromatography samples
  • Manufactured from amber 203 borosilicate glass for chemical resistance and light protection
  • Features an aluminum seal finish; supplied without closure, making them lighter than molded bottles
  • Enhanced by the most popular GPI aluminum seal finishes, offering a variety of choices
  • Ideal for packaging and storage where injectable and parenteral solution applications require utmost purity

Specifications

Color Amber
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass (Type 1)
Pre-slit No
Sterility Non-sterile
Volume (Metric) 2 mL
For Use With (Application) Freeze drying applications
Material (Seal) Aluminum
Volume (English) 0.5 dr.
Quantity 1440/Cs.
Marking Spot No
Autoclavable Autoclavable
Certifications/Compliance ASTM E438, Type I
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less