missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ KnockOut™ Serum Replacement
GREENER_CHOICE

Numéro de catalogue. p-7076000
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
10828010 100 mL
10828028 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 10828010 Fournisseur Gibco™ Code fournisseur 10828010
Il en reste null

KnockOut Serum Replacement (KnockOut SR) is a more defined, FBS-free medium supplement that supports the growth of pluripotent stem cells (PSCs) cultured on fibroblast feeder cells.

Gibco KnockOut Serum Replacement (KnockOut SR) is a more defined, FBS-free medium supplement that supports the growth of pluripotent stem cells (PSCs) cultured on fibroblast feeder cells. Designed to replace fetal bovine serum (FBS) in existing protocols, this serum replacement has been trusted by many for almost 20 years. Providing the documents required for regulatory review, KnockOut SR reduces your burden in qualifying reagents when transitioning from research applications to clinical applications.

With KnockOut Serum Replacement, you can:

  • Culture feeder-dependent PSCs with confidence
  • Rely on the improved performance of a qualified, FBS-free formulation
  • Enjoy the peace of mind of having a more stable price and supply compared to FBS

Culture PSCs with confidence

KnockOut SR has been cited in more than 2000 publications. It has been shown to support feeder-dependent culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs). It has also been shown to support reprogramming, embryoid body (EB) formation, in vitro differentiation, cryopreservation, ESC derivation, and many other applications.

Rely on improved performance

KnockOut SR has more a defined formulation compared to FBS, which contains many undefined and variable components. Unlike most FBS products, each lot is tested for its ability to support PSCs. Because of this, culturing PSCs with KnockOut SR leads to less differentiation and more compact colony morphology compared to FBS.

Enjoy the peace of mind of having stable price and supply

KnockOut SR is not affected by the limited supply and fluctuating prices often seen with FBS. This makes it easier to plan your budget and buy what you need, especially since it is more affordable than most FBS products.

Spécifications

Classification Serum-Free
Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Culture Type Mammalian Cell Culture
Species Serum-Free
Cell Type Stem Cells (Embryonic)
Form Frozen
Serum Level Serum-Free
Product Type Serum Replacement
Quantity 100 mL
Tested For Growth Performance Assay
Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
For Use With (Application) Clinical Research, Translational Research
Purity or Quality Grade ES Cell-Qualified
Product Line KnockOut
pH 6 to 8
Shipping Condition Dry Ice
Afficher plus de résultats Afficher moins de résultats

For research use only. Not for use in diagnostic procedures.