KLH-Separopore 4B Affinity matrix with immobilized KLH for the purification of antibodies generated against a peptide-KLH conjugated antigens. It allows removal of antobodies against KLH portion of the antigen; Supplied as a suspension i...
Numéro de catalogue.
50197700
Fournisseur
BioworldCode fournisseur
201810163
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
KLH-Separopore 4B Affinity matrix with immobilized KLH for the purification of antibodies generated against a peptide-KLH conjugated antigens. It allows removal of antobodies against KLH portion of the antigen; Supplied as a suspension i...