missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KL 001
Description
Cryptochrome protein (CRY) stabilizer; blocks proteasome-mediated degradation of CRY1 and CRY2. Lengthens circadian period in mouse NIH-3T3 fibroblasts without altering casein kinase activity. Represses glucagon-induced hepatic gluconeogenesis through the induction of Pck1 and G6pc expression.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 398.48 |
| Purity | >98% |
| Molecular Weight (g/mol) | 398.477 |
| Chemical Name or Material | KL 001 |
Safety and Handling
| Recommended Storage | Store at +4°C |