accessibility menu, dialog, popup

UserName

H3CSOONONHO

CAS RN 309928-48-1

IUPAC Name: N-[3-(9H-carbazol-9-yl)-2-hydroxypropyl]-N-[(furan-2-yl)methyl]methanesulfonamide
Synonyms: N-[3-(carbazol-9-yl)-2-hydroxypropyl]-N-(furan-2-ylmethyl)methanesulfonamide
Molecular Weight (g/mol): 398.48
Molecular Formula: C21H22N2O4S
InChi Key: OQAFDLPAPSSOHY-UHFFFAOYNA-N
SMILES: CS(=O)(=O)N(CC(O)CN1C2=C(C=CC=C2)C2=C1C=CC=C2)CC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results