Learn More
BUCHI Kjeldahl Sample Tubes
Sample tubes for standard Kjeldahl are suitable for most applications.
Supplier:  BUCHI 026128
 Afficher plus de versions de ce produit
Numéro de catalogue. 05405647
Il en reste null
Description
Description
  • Precision joint: tightness, avoid fume leaks
  • Constricted condensation zone; no splashing
  • Wider diameter; reduced foaming
  • Thicker glass wall, longer lifetime
Spécifications
Identifiants chimiques
Sample Tube
Spécifications
Large Volume Samples
Sample Tube
500 mL
Each
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS