Learn More
Thermo Scientific™ Kisspeptin-10
For cellular and molecular biology applications
Supplier:  Thermo Scientific™ J65487MCR
 View more versions of this product
Catalog No. AAJ65487MCR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chemical Identifiers
Kisspeptin-10
-30°C to -10°C
Powder
RUO - research use only
1302.44
Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH2
Metastin (45-54), amide, human
HPLC
Specifications
HPLC
-30°C to -10°C
1302.44
1 mg
Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH2
Metastin (45-54), amide, human
Powder
Kisspeptin-10
RUO - research use only
Product Suggestions
Product Suggestions