Learn More
Thermo Scientific™ KingFisher 96 Deep-well, (for KF Flex/Duo Prime only)
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.
$748.61 - $1,681.04
| Numéro de catalogue | Numéro du manufacturier. | Product Type | For Use With (Equipment) | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Product Type | For Use With (Equipment) | Prix | Quantity & Availability | ||||
|
22387031
|
Thermo Scientific™
95040450 |
Deep-well Plate | KF Duo Prime, Flex and Presto |
paquet de 50 chaque for $748.61
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
22387032
|
Thermo Scientific™
95040460 |
Deep-well Plate | KF Duo Prime, Flex and Presto |
chaque for $833.80
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
14387349
|
Thermo Scientific™
95040450B |
Deep-well Plate | KingFisher Apex |
paquet for $753.65
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
14387350
|
Thermo Scientific™
95040460B |
Deep-well Plate | KingFisher Apex |
paquet for $907.81
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
FER97002820
|
Thermo Scientific™
97002820 |
Tip Comb and Deep-well Plate | KingFisher Flex and Presto |
chaque for $1,346.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
14387358
|
Thermo Scientific™
97002820B |
Tip Comb and Deep-well Plate | KingFisher Apex |
paquet for $1,681.04
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Spécifications
| 96 |
For research use only. Not for use in diagnostic procedures.