Learn More
DWK Life Sciences KIMBLE™ Replacement Stopcocks for KIMAX™ Microburet with Three-Way PTFE Stopcock
Description
- Replacement three-way PTFE stopcock plug for KIMAX™ microburets and reservoir-fill burets and pipets requiring standard taper PTFE stopcock plugs
- Chemically resistant and self-lubricating PTFE construction
- Plug size: 12.9/44 mm
- Each plug furnished with a nut, rubber washer, and HDPE washer
Specifications
Specifications
| Type | Replacement Stopcock |
| Plug Bore | 2 mm |
| For Use With (Application) | Microburet |
| For Use With (Equipment) | Kimax Microburet |
| Includes | Nut, rubber and HPDE washers |
| Plug Material | PTFE |
| Connectors | 3-way |