Learn More
DWK Life Sciences™ Kimble™ Polyacetal Standard Taper Joint Clamp
The Kimble™ Polyacetal Standard Taper Joint Clamp is used to secure Standard Taper joints. This positive closure clamp is made of tough, color-coded plastic.
Supplier:  DWK Life Sciences™ 6753000034
 Afficher plus de versions de ce produit
Numéro de catalogue. 01183035
Il en reste null
Description
Description
  • Manufactured from polyacetal which is characterized by its high strength, hardness and rigidity to -40°C, and offers good overall chemical resistance
  • Joint clamps are conveniently colour coded to help with size identification
  • The clamp will not scratch glass and can be autoclaved to 200°C
Spécifications
Identifiants chimiques
Polyacetal Standard Taper Joint Clamp
Polyacetal
Spécifications
Polyacetal Standard Taper Joint Clamp
Polyacetal
34/28, 34/45
Orange
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS