Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners
Cap provides good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents.
Supplier:  DWK Life Sciences 75201G33400
 View more versions of this product
Catalog No. 13757117
Only null left
Alert:
Not recommended for hydrocarbons or bleaches.
Description
Description
  • Polyvinyl film is adhered to 0.035-in. pulpboard
  • Color: black
  • Low cost cap for general purpose usage
  • Not recommended for hydrocarbons or bleaches
Specifications
Chemical Identifiers
Black
144 Cs.
Bottle
Phenolic Resin
Phenolic Cap
Pulp/Vinyl
Specifications
Black
144 Cs.
Phenolic Cap
Pulp/Vinyl
Phenolic Resin
33-400
Bottle
Product Suggestions
Product Suggestions