missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners

Catalog No. 13757126
Change view
Click to view available options
Neck Finish:
20-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
70-400
89-400
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
13757126 89-400
13757116 28-400
13757113 20-400
13757118 38-400
13757117 33-400
13757115 24-400
13757119 43-400
13757125 70-400
13757123 58-400
13757120 45-400
13757121 48-400
13757122 53-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Not recommended for hydrocarbons or bleaches.
Catalog No. 13757126 Supplier DWK Life Sciences Supplier No. 75201G89400
Not recommended for hydrocarbons or bleaches.

Cap provides good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents.

  • Polyvinyl film is adhered to 0.035-in. pulpboard
  • Color: black
  • Low cost cap for general purpose usage
  • Not recommended for hydrocarbons or bleaches

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 89-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner Pulp/Vinyl