missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Phenolic Caps with PTFE-Faced LDPE Foam Liners
Offers excellent chemical and solvent resistance
$296.25 - $1,938.96
Specifications
| Color | Black |
|---|---|
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Product Type | Phenolic Cap |
| For Use With (Equipment) | Bottle |
Alert:
Poor for organic amines.
| Catalog Number | Mfr. No. | Neck Finish | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | Price | Quantity & Availability | |||||
|
13757175
|
DWK Life Sciences
75206G20400 |
20-400 |
Case of 144 Each for $296.25
|
|
|||||
|
13757178
|
DWK Life Sciences
75206G28400 |
28-400 |
Case of 144 Each for $326.25
|
|
|||||
|
13757179
|
DWK Life Sciences
75206G33400 |
33-400 |
Case of 144 Each for $416.00
|
|
|||||
|
13757180
|
DWK Life Sciences
75206G38400 |
38-400 |
Case of 144 Each for $507.79
|
|
|||||
|
13757176
|
DWK Life Sciences
75206G22400 |
22-400 |
Case of 144 Each for $325.64
|
|
|||||
|
13757182
|
DWK Life Sciences
75206G45400 |
45-400 |
Case of 144 Each for $459.87
|
|
|||||
|
13757181
|
DWK Life Sciences
75206G43400 |
43-400 |
Case of 144 Each for $404.44
|
|
|||||
|
13757183
|
DWK Life Sciences
75206G48400 |
48-400 |
Case of 144 Each for $750.64
|
|
|||||
|
13757184
|
DWK Life Sciences
75206G53400 |
53-400 |
Case of 144 Each for $886.93
|
|
|||||
|
13757186
|
DWK Life Sciences
75206G63400 |
63-400 |
Case of 144 Each for $930.95
|
|
|||||
|
13757187
|
DWK Life Sciences
75206G70400 |
70-400 |
Case of 144 Each for $1,390.36
|
|
|||||
|
13757188
|
DWK Life Sciences
75206G89400 |
89-400 |
Case of 144 Each for $1,938.96
|
|
|||||
|
13757185
|
DWK Life Sciences
75206G58400 |
58-400 |
Case of 144 Each for $1,104.32
|
|
|||||
Description
- Excellent solvent resistance
- General purpose
- Liner provides compressible foam for sealability
All caps/liners should be tested to ensure leak and compatibility performance with contents.
Specifications
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| PTFE/LDPE Foam |