missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with PTFE-Faced LDPE Foam Liners

Catalog No. 13757175
Change view
Click to view available options
Neck Finish:
20-400
22-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
13757175 20-400
13757178 28-400
13757179 33-400
13757180 38-400
13757176 22-400
13757182 45-400
13757181 43-400
13757183 48-400
13757184 53-400
13757186 63-400
13757187 70-400
13757188 89-400
13757185 58-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Poor for organic amines.
Catalog No. 13757175 Supplier DWK Life Sciences Supplier No. 75206G20400
Poor for organic amines.

Offers excellent chemical and solvent resistance

  • General purpose phenolic screw cap with foam liner
  • Color: black
  • Liner material: 0.005-in. PTFE-faced 0.050-in. LDPE foam
  • Poor for organic amines
  • Compressible foam for sealability

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 20-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner PTFE/LDPE Foam