Learn More
DWK Life Sciences Kimble™ Phenolic Caps with PTFE-Faced LDPE Foam Liners
Offers excellent chemical and solvent resistance
Supplier: DWK Life Sciences 75206G70400
Alert:
Poor for organic amines.
Description
- General purpose phenolic screw cap with foam liner
- Color: black
- Liner material: 0.005-in. PTFE-faced 0.050-in. LDPE foam
- Poor for organic amines
- Compressible foam for sealability
Chemical Identifiers
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| PTFE/LDPE Foam |
Specifications
| Black | |
| 144 Cs. | |
| Phenolic Cap | |
| PTFE/LDPE Foam |
| Phenolic Resin | |
| 70-400 | |
| Bottle |