Learn More
DWK Life Sciences Kimble™ Phenolic Caps with PTFE-Faced LDPE Foam Liners
Offers excellent chemical and solvent resistance
Supplier:  DWK Life Sciences 75206G63400
 View more versions of this product
Catalog No. 13757186
Only null left
Alert:
Poor for organic amines.
Description
Description
  • General purpose phenolic screw cap with foam liner
  • Color: black
  • Liner material: 0.005-in. PTFE-faced 0.050-in. LDPE foam
  • Poor for organic amines
  • Compressible foam for sealability
Specifications
Chemical Identifiers
Black
144 Cs.
Bottle
Phenolic Resin
Phenolic Cap
PTFE/LDPE Foam
Specifications
Black
144 Cs.
Phenolic Cap
PTFE/LDPE Foam
Phenolic Resin
63-400
Bottle
Product Suggestions
Product Suggestions