missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Solvent Filtration Assemblies
Description
Includes
- Solvent pickup adapter
- Coarse porosity fritted glass filter support with No. 2 vacuum hose connection
- Anodized aluminum clamp
- Filter flask
- Draw solvent directly from reagent bottle through 47mm filter and into filtration flask, eliminating the need to pour HPLC solvents into funnels
- After filtration, stopcock on adapter can be turned off for vacuum degassing
- Solvent pickup adapter available separately to convert funnel assemblies
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
- Joints are 40/35
Specifications
Specifications
| Disposable | Reusable |
| Material | Glass |
| Sterility | Non-sterile |
| Type | Flask |
| Volume (Metric) | 4 L |
| Width (English) | 8.0625 in. |
| Width (Metric) | 20.5 cm |
| Height (English) | 20.66 in. |
| Height (Metric) | 52.5 cm |
| For Use With (Equipment) | Filtration Assembly and vacuum pump |