Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached White Rubber Lined Caps
Packaged in corrugated trays with partitions
 View more versions of this product
Catalog No. 14957213
Only null left
Description
Description
  • Small case quantities
  • Manufactured from clear 33 expansion borosilicate glass
  • Attached closed top black phenolic closures
Specifications
Chemical Identifiers
A
Clear
Screw
Screw Cap (Closed-top)
Rubber
Borosilicate Glass
Phenolic
Specifications
A
Clear
Borosilicate Glass
Screw
Phenolic
200/Cs.
Rubber
15 mm
2 mL
0.5 dr.
Screw Cap (Closed-top)
28 mm
Product Suggestions
Product Suggestions