Learn More
DWK Life Sciences Kimble™ Clear Sample Vial with Rubber Cap
Clear screw thread sample vial
Supplier:  DWK Life Sciences 60910D3
 Afficher plus de versions de ce produit
Numéro de catalogue. 12141498
Il en reste null
Description
Description
  • Closed-top black phenolic closure with rubber liner, unattached
Spécifications
Identifiants chimiques
Black
Black phenolic caps and rubber liners
Sample Vial
Clear
Phenolic with Rubber Liner
Spécifications
Black
19 mm
Black phenolic caps and rubber liners
Phenolic with Rubber Liner
65 mm
Clear
12 mL
15-425
Type I, 51 expansion borosilicate glass tubing
Sample Vial
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS