missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Clear Glass Straight-Sided Jars, Tall, Without Cap

Wide-mouth design

$131.07 - $175.86

Specifications

Color Clear
For Use With (Application) Storage
Includes Cap
Material Soda Lime Glass
Packaging Bulk
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
1377484
View Documents Promotion Details
DWK Life Sciences
5510448B
125 mL 48-400 mm
Case of 24 Each for $131.07
Only null left
 
1377485
Accessories View Documents Promotion Details
DWK Life Sciences
5510858B
250 mL 58-400 mm
Case of 24 Each for $160.00
Only null left
 
1377486
View Documents Promotion Details
DWK Life Sciences
5511670B
500 mL 70-400 mm
Case of 12 Each for $175.86
Only null left
 
Description

Description

  • Clear glass for easy viewing of contents
  • Type III soda-lime glass
  • Caps sold separately

All 32-ounce jars in this product family are packed in corrugated cartons with divider cells.

Specifications

Specifications

Clear
Cap
Bulk
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Cap
Storage
Soda Lime Glass
Clear
Wide
Straight-sided Jar
Videos
SDS
Documents

Documents