missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Clear Glass Straight-Sided Jars, Tall, Without Cap

Wide-mouth design

Supplier:  DWK Life Sciences 5510858B

 View more versions of this product

Catalog No. 1377485


Only null left

Description

Description

  • Clear glass for easy viewing of contents
  • Type III soda-lime glass
  • Caps sold separately

All 32-ounce jars in this product family are packed in corrugated cartons with divider cells.

Specifications

Chemical Identifiers

Clear
Cap
Bulk
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Cap
Storage
Soda Lime Glass
Clear
Wide
Straight-sided Jar

Specifications

8 oz., 250 mL
8 oz.
2.44 in.
Cap
Bulk
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Straight-sided Jar
58-400 mm
Cap
250 mL
Clear
Storage
Soda Lime Glass
Clear
Wide
127 mm
5 in.
62 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions