missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Clear Glass Straight-Sided Jars with 14B White-Rubber-Lined Caps

Wide-mouth design for efficient addition and removal of contents

$200.21 - $1,285.36

Specifications

Color Clear
For Use With (Application) Storage
Material Soda Lime Glass
Closure Color Black
Clarity Clear
View More Specs

Products 6
Catalog Number Mfr. No. Capacity (Metric) Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Packaging Price Quantity & Availability  
13756758
View Documents Promotion Details
DWK Life Sciences
5410253V24
60 mL Convenience
Case of 24 Each for $200.21
Only null left
 
13756765
View Documents Promotion Details
DWK Life Sciences
5410458C24
125 mL Bulk
Case of 144 Each for $1,285.36
Only null left
 
13756759
View Documents Promotion Details
DWK Life Sciences
5410458V24
125 mL Convenience
Case of 24 Each for $212.75
Only null left
 
13756767
missing translation for 'viewDocuments' Promotion Details
DWK Life Sciences
5410870C24
250 mL Bulk
Case of 24 Each for $411.32
Only null left
 
13756768
missing translation for 'viewDocuments' Promotion Details
DWK Life Sciences
5411689C24
500 mL Bulk
Case of 12 Each for $260.75
Only null left
 
13756769
missing translation for 'viewDocuments' Promotion Details
DWK Life Sciences
5413289C24
1000 mL Bulk
Case of 12 Each for $249.54
Only null left
 
Description

Description

All 32-ounce jars are packed in corrugated cartons with divider cells.

Specifications

Specifications

Clear
Soda Lime Glass
Clear
Wide
Straight-sided Jar
Phenolic
Storage
Black
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Rubber
Screw Cap
Videos
SDS
Documents

Documents