Learn More
DWK Life Sciences Kimble™ Clear Boston Rounds without Caps
Narrow-mouth design provides more controlled pouring than wide-mouth.
Supplier:  DWK Life Sciences 5110824B
 View more versions of this product
Catalog No. 13756941
Only null left
Description
Description
  • High quality, economical narrow mouth bottles are manufactured from clear Type III soda-lime glass to ensure easy viewing of the contents.
Specifications
Chemical Identifiers
Clear
Boston Round Bottle
Specifications
8 oz., 250 mL
8 oz.
2.36 in.
136 mm
5.3 in.
60 mm
250 mL
Clear
Soda Lime Glass
Boston Round Bottle
24-400 mm
Product Suggestions
Product Suggestions