Learn More
DWK Life Sciences Kimble™ Micro Hematocrit Capillary Tubes
Kimble™ micro-hematocrit tubes are used to measure the volume percentage of red blood cells in blood.
Supplier:  DWK Life Sciences 40C505
 Afficher plus de versions de ce produit
Numéro de catalogue. 14955400
Il en reste null
Description
Description
Kimble™ micro-hematocrit tubes are used to measure the volume percentage of red blood cells in blood.
  • Packed in plastic vials with reclosable snap-lock caps to maintain cleanliness
  • Ends may be sealed by flame or sealing clay
  • Heparinized capillary tubes are designed to prevent blood clotting
  • Ammonium heparin does not interfere with sodium identification
  • Tubes are manufactured from 90 expansion soda lime glass conforming to USP Type III requirements
Spécifications
Identifiants chimiques
Soda Lime Glass
Used to measure the volume percentage of red Blood cells in Blood
USP type III
Snap-lock
75 mm
60 mm
Spécifications
Soda Lime Glass
0.5 mm
75 mm
60 mm
Snap-lock
Used to measure the volume percentage of red Blood cells in Blood
USP type III
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS