Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
$372.59 - $752.46
Specifications
For Use With (Application) Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
View More Specs
Products 10
Catalog Number Mfr. No. Capacity (Metric) Material Includes Dnase Free Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Material Includes Dnase Free Price Quantity & Availability  
K7495150000
Accessories View Documents Promotion Details
DWK Life Sciences
7495150000
1.5 mL CTFE/Stainless Steel w/o Microtubes No
Each for $393.50
Only null left
K7495160500
View Documents Promotion Details
DWK Life Sciences
7495160500
0.5 mL CTFE/Stainless Steel w/o Microtubes No
Each for $530.39
Only null left
K7495200000
Accessories View Documents Promotion Details
DWK Life Sciences
7495200000
1.5 mL Polypropylene 100 Microtubes No
Case of 100 Each for $586.29
Only null left
K7495200500
Accessories View Documents Promotion Details
DWK Life Sciences
7495200500
0.5 mL Polypropylene 100 Microtubes No
Case of 100 Each for $527.79
Only null left
K7495210590
Accessories View Documents Promotion Details
DWK Life Sciences
7495210590
0.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free
Case of 100 Each for $586.86
Only null left
K7495200090
Accessories View Documents Promotion Details
DWK Life Sciences
7495200090
1.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free
Case for $752.46
Only null left
K7495200590
Accessories View Documents Promotion Details
DWK Life Sciences
7495200590
0.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free
Case for $706.57
Only null left
K7495211590
Accessories View Documents Promotion Details
DWK Life Sciences
7495211590
1.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free
Case of 100 Each for $372.59
Only null left
K7495210500
Accessories View Documents Promotion Details
DWK Life Sciences
7495210500
0.5 mL Polypropylene w/o Microtubes No
Case of 100 Each for $426.25
Only null left
K7495211500
Accessories View Documents Promotion Details
DWK Life Sciences
7495211500
1.5 mL Polypropylene w/o Microtubes No
Pack of 100 Each for $464.96
Only null left
Description
Description
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Disposable Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size
Specifications
Specifications
Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes