Learn More
DWK Life Sciences KIMBLE™ KONTES™ MIDI-VAP 4000 Distillation Glassware Set
Description
Glassware sets for both ammonia and phenol distillation system
- Glassware sets for the Kimble™ Kontes™ MIDI-VAP 4000 Distillation Apparatus
- Supports ammonia, phenol, and cyanide distillation applications
- Includes glassware sets, tubing kits, reaction tubes, distilling heads, caps, septa, sealing rings, quick disconnects, and manifold heater
- Borosilicate glass components designed for reduced sample volume semi-automatic colorimetry methods
- Compatible with drinking, ground, surface, and saline water analysis as well as domestic and industrial waste testing
Specifications
Specifications
| For Use With (Application) | Distillation Systems |
| Type | Cyanide Distillation System |
| For Use With (Equipment) | MIDI-VAP 4000 |
| Quantity | 5/Pk. |