missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Center Well for Incubation Flask

Catalog No. K8823200000
Change view
Click to view available options
No. per Pack:
144
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
K8823200000 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K8823200000 Supplier DWK Life Sciences Supplier No. 8823200000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The KIMBLE™ KONTES™ Center Well for Incubation Flask is a disposable polypropylene accessory for the KONTES™ sidearm incubation flask. Length 70 mm, diameter 10 mm. The center well may be cut off when transferred to a liquid scintillation counting vial.

  • Disposable polypropylene center well designed for use with the Kontes™ sidearm incubation flask
  • Inserted through the off-center hole in the rubber top stopper, which provides a double seal
  • Center well may be cut off when transferred to a liquid scintillation counting vial
  • Sidearm flask system enables syringe needle entry without disturbing established incubation conditions
  • Suitable for metabolic studies including measurement of C14O2 yields, in vitro incubations, insulin bio-assays, persulfate oxidations, and distillation of volatile acids or bases
  • Compatible flask manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Sold separately in case quantities of 144

Specifications

Diameter (English) Outer 0.39 in.
Material Polypropylene
For Use With (Application) For metabolic studies of biological materials
For Use With (Equipment) For incubation flask
No. per Pack 144
Product Type Center well
Diameter (Metric) Outer 1 cm