Learn More
DWK Life Sciences KIMBLE™ KONTES™ Hi-Vac Straight Low Hold Up Valves without Tip O-Ring
Catalog No. K8266010008
Change view
Click to view available options
Plug Stopcock No:
4
8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Stopcock No
K8266010008 8
K8266010004 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K8266010008 Supplier DWK Life Sciences Supplier No. 8266010008
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KIMBLE™ KONTES™ Hi-Vac Straight Low Hold Up Valves without Tip O-Ring feature bent sidearms to minimize liquid holdup. The PTFE plug provides precise flow control for both liquids and gases, suitable for high-vacuum laboratory applications.
  • Bent sidearms minimize liquid holdup compared to conventional straight valves
  • PTFE plug provides precise flow control for both liquids and gases
  • Designed for high-vacuum laboratory applications
  • Available in multiple bore sizes to suit different application requirements
  • Manufactured from 33-expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Actuation Manual
Volume (Metric) Sample Size 8
For Use With (Equipment) HI-Vac Straight Low Hold Up Valves
Material PTFE
Plug Stopcock No 8
Type Metering High Vacuum Valve Plug
Stator Material PTFE/CTFE