missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Buret with PTFE-Plug Stopcocks Replacement Glass Body

Catalog No. 0370021C
Change view
Click to view available options
Capacity (Metric):
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0370021C 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0370021C Supplier DWK Life Sciences Supplier No. 17026G50

With Lubri-Flo™ stopcock barrel.

  • Without PTFE plug
  • Capacity: 50mL

Specifications

Capacity (Metric) 50 mL
Material Soda Lime Glass
Includes Lubri-Flo Stopcock Barrel
Stopcock Material PTFE
Type Bore Burette Barrel
Class Class B