missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Black Phenolic Screw Caps
Description
Bond between liner and cap retains adhesive properties even after repeated autoclaving
With PTFE-faced rubber liner
- Identified in ASTM™ specification E 982-94, Class B requirements
- Has fewer threads than regular GPI caps for easier application and removal
- Not recommended for use with standard GPI finishes usually found on blown-molded bottles
- Identified in ASTM specification E 982-94, Class A requirements
Spécifications
Spécifications
| Color | Black |
| Height (Metric) With Closure | 26mm |
| Material | Phenolic Resin |
| No. per Case | 150 Cs. |
| Neck Finish | 38-430 |
| Certifications/Compliance | ASTM E982, Class A |
| For Use With (Equipment) | Bottle |
| Closure Liner | 14B Rubber |
| Autoclavable | Autoclavable |