Learn More
DWK Life Sciences Kimble™ KIMAX™ Media Bottles — Ungraduated
Designed for easy pouring and handling
$357.50 - $967.14
Specifications
Color White
Material Borosilicate Glass
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
06-421-1
Accessories View Documents
DWK Life Sciences
61100125
125 mL 33-430 mm
Case of 48 Each for $439.68
Only null left
06-421-3
Accessories View Documents
DWK Life Sciences
61100250
250 mL 33-430 mm
Case of 48 Each for $967.14
Only null left
06-421-2
Accessories View Documents
DWK Life Sciences
61100500
500 mL 33-430 mm
Case of 24 Each for $543.34
Only null left
06-421-6
Accessories View Documents
DWK Life Sciences
611001000
1000 mL 33-430 mm
Case of 12 Each for $357.50
Only null left
06-421-5
Accessories View Documents
DWK Life Sciences
61100A1000
1000 mL 38-430 mm
Case of 12 Each for $392.42
Only null left
Description
Description
  • Available with (Cat. No. 06-421-7) or without graduations
  • 1000mL size features finger grips molded into the side wall and is available with two GPI thread sizes
  • All bottles have screw-thread finish
  • Without closures
Specifications
Specifications
White
Borosilicate Glass