Learn More
DWK Life Sciences Kimble™ KIMAX™ Borosilicate Petri Dish Set
Shallow glass dishes used for culturing bacteria and other general laboratory uses. Designed to meet Federal Specification NNN-D-1478.
$1,135.93 - $1,274.57
Specifications
Bottom Shape Flat
Color Clear
Material Borosilicate Glass
Shape Round
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Price Quantity & Availability  
02555130
View Documents Promotion Details
DWK Life Sciences
2306110015
15 mm
Case of 36 Each for $1,274.57
Only null left
02555131
View Documents Promotion Details
DWK Life Sciences
2306110020
20 mm
Case of 34 Each for $1,135.93
Only null left
Description
Description
Shallow glass dishes used for culturing bacteria and other general laboratory uses. Designed to meet Federal Specification NNN-D-1478.
  • Kimble Kimax dishes remain clear after repeated use in wet or dry sterilization cycles
  • Reinforced beaded edges resist mechanical breakage and help in centering the bottom inside the cover
  • Covers have a fused-on bright red ceramic enamel marking spot and inscription, including “Cover,” Bottoms have a white ceramic enamel inscription, including “Bottom”
  • Combination of different colors and wording permits quick and easy identification of the parts when sorting, selecting and assembling pairs
  • Bottom has a vertical arrow on the side to precisely locate serial dilution starting points
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Flat
Borosilicate Glass
Non-sterile
100 mm
Clear
Round
Petri Dish Set