missing translation for 'onlineSavingsMsg'
Learn More

Khellin 95.0+%, TCI America™

Catalog No. K00395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00395G 5 g
K003925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00395G Supplier TCI America Supplier No. K00395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82-02-0
Molecular Formula C14H12O5
Molecular Weight (g/mol) 260.245
MDL Number MFCD00005007
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
Synonym khellin, visammin, amicardine, methafrone, coronin, amikellin, ammipuran, ammivisnagen, benecardin, corafurone
PubChem CID 3828
ChEBI CHEBI:6133
IUPAC Name 4,9-dimethoxy-7-methylfuro[3,2-g]chromen-5-one
SMILES CC1=CC(=O)C2=C(C3=C(C(=C2O1)OC)OC=C3)OC

Specifications

Melting Point 152°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 260.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Khellin
RTECSNumber LV1050000
TSCA No