missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Reagents

Numéro de catalogue. 01337204
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01337204 1 L
01337203 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 01337204 Fournisseur Labchem Inc Code fournisseur C22220001C
Il en reste null

Kerosene, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 8008-20-6
Molecular Formula C12H26-C15H32
Synonym Paraffin oil

Spécifications

Boiling Range 175°C to 325°C
Melting Point -48°C to -26°C
Color Red
Boiling Point 175°C
Melting Range -48°C to -26°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.767
Grade Technical
Physical Form Liquid
Chemical Name or Material Kerosene
Afficher plus de résultats Afficher moins de résultats