missing translation for 'onlineSavingsMsg'
Learn More

Kenpaullone 97.0+%, TCI America™

Catalog No. K005250MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K005250MG 50 mg
K005210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K005250MG Supplier TCI America Supplier No. K005250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142273-20-9
Molecular Formula C16H11BrN2O
Molecular Weight (g/mol) 327.181
MDL Number MFCD02683595
InChI Key QQUXFYAWXPMDOE-UHFFFAOYSA-N
Synonym kenpaullone, 9-bromopaullone, 9-bromo-7,12-dihydroindolo 3,2-d 1 benzazepin-6 5h-one, indolo 3,2-d 1 benzazepin-6 5h-one,9-bromo-7,12-dihydro, 9-bromo-7,12-dihydro-indolo 3,2-d 1 benzazepin-6 5h-one, 9-bromo-7,12-dihydro-5h-indolo 3,2-d 1 benzazepin-6-one, 9-bromo-7,12-dihydro-indolo-3,2-d-1 benzazepin-6 5h-one, 9-bromo-7,12-dihydrobenzo 2,3 azepino 4,5-b indol-6 5h-one, indolo 3,2-d 1 benzazepin-6 5h-one, 9-bromo-7,12-dihydro
PubChem CID 3820
IUPAC Name 9-bromo-7,12-dihydro-5H-indolo[3,2-d][1]benzazepin-6-one
SMILES C1C2=C(C3=CC=CC=C3NC1=O)NC4=C2C=C(C=C4)Br

Specifications

Quantity 50 mg
Formula Weight 327.18
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Kenpaullone
TSCA No
Recommended Storage Freezer