missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keller's Reagent (Etchant), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Keller's Reagent |
| Color | Colorless |
| CAS | 7732-18-5, 7697-37-2, 7647-01-0, 7664-39-3 |
| CAS Min % | 92.0, 3.62, 1.71, 1.12 |
| CAS Max % | 95.0, 3.77, 1.78, 1.16 |
| Synonym | Etchant |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | Acidic |
| Quantity | 1 L |
| Show More |